Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C12orf29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009894.2 | NP_001009894.2 | 6 | 930 | ||
GenBank mRNA | AK095477.1 | - | 6 | 930 | ||
AK311153.1 | - | 4 | 572 | |||
AK311222.1 | - | 5 | 798 | |||
AL137488.1 | - | 5 | 2838 | |||
BC035136.2 | - | 5 | 767 | |||
BC058014.1 | - | 5 | 795 | |||
BC092422.1 | - | 6 | 872 | 30386540 | ||
BX537868.1 | - | 6 | 902 | |||
BX640968.1 | - | 6 | 905 | |||
BX648026.1 | - | 5 | 766 | |||
CR936776.1 | - | 6 | 919 |