Search
Search

LINC01278| Gene Symbol: LINC01278 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015353.1 | 4 | 2853 | |||
| GenBank mRNA | AK001756.1 | - | 4 | 2853 | ||
| AK023798.1 | - | 1 | 3929 | |||
| AK129670.1 | - | 1 | 1856 | |||
| AK130294.1 | - | 1 | 5056 | |||
| AL080202.1 | - | 3 | 2736 | |||
| BC024701.1 | - | 1 | 582 | |||
| BC048208.1 | - | 1 | 1841 | |||