Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other FHAD1 SIRNA Assays ›
Gene Symbol:
FHAD1
Gene Name:
forkhead associated phosphopeptide binding domain 1
Gene Aliases:
-
Chromosome Location:
Chr.1: 15236559 - 15400283 on Build GRCh38
UniGene ID:
Hs.659997
Species:
Human
siRNA ID
HSS150969
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: FHAD1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_052929.1
NP_443161.1
21
2913
XM_011540575.2
XP_011538877.1
22
3055
XM_011540576.2
XP_011538878.1
22
3056
XM_011540577.2
XP_011538879.1
22
3017
XM_011540580.2
XP_011538882.1
22
3054
XM_011540581.2
XP_011538883.1
22
3054
XM_011540582.2
XP_011538884.1
22
3054
XM_011540584.2
XP_011538886.1
22
3055
XM_011540588.2
XP_011538890.1
22
3046
XM_011540589.2
XP_011538891.1
21
2947
XM_011540590.2
XP_011538892.1
22
3047
XM_011540592.2
XP_011538894.1
21
2756
XM_011540593.2
XP_011538895.1
21
2943
XM_011540595.2
XP_011538897.1
19
2399
XM_011540596.2
XP_011538898.1
16
1959
XM_011540597.2
XP_011538899.1
22
3056
XM_017000198.1
XP_016855687.1
22
3056
XM_017000199.1
XP_016855688.1
22
3056
XM_017000200.1
XP_016855689.1
22
3055
XM_017000201.1
XP_016855690.1
22
3055
XM_017000202.1
XP_016855691.1
21
2948
XM_017000203.1
XP_016855692.1
22
3052
XM_017000204.1
XP_016855693.1
22
3051
XM_017000205.1
XP_016855694.1
20
2861
XM_017000206.1
XP_016855695.1
22
3051
XM_017000207.1
XP_016855696.1
21
2820
XM_017000208.1
XP_016855697.1
22
3046
XM_017000209.1
XP_016855698.1
22
3050
XM_017000210.1
XP_016855699.1
22
3046
XM_017000211.1
XP_016855700.1
21
2946
XM_017000212.1
XP_016855701.1
22
3045
XM_017000213.1
XP_016855702.1
22
3055
XM_017000214.1
XP_016855703.1
22
3056
GenBank mRNA
AB067524.1
-
6
765
AK093883.1
-
4
480
AK296920.1
-
5
588
AL833342.1
-
11
1312
Back To Top
More Information
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5c7f9f6c68-zgjsk:80/10.239.161.116:80.
git-commit: b16971a8df2afcd8df684aafbc9bf1e9f45e42d4
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.6.0-2024.04.15-1.0