Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCTD12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138444.3 | NP_612453.1 | 1 | 486 | ||
GenBank mRNA | AF359381.1 | - | 1 | 471 | ||
AK128363.1 | - | 1 | 489 | |||
BC013764.1 | - | 1 | 316 | 4521096 |