Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPACA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173847.4 | NP_776246.1 | 2, 3 | 410 | ||
GenBank mRNA | AF216311.1 | - | 2, 3 | 410 | ||
AY358653.1 | - | 3, 4 | 537 | |||
BC100887.1 | - | 2, 3 | 368 | 40003365 | ||
BF511575.1 | - | 3, 4 | 424 |