Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other EEF1D SIRNA Assays ›
Gene Symbol:
EEF1D
ANKRD19P
Gene Name:
eukaryotic translation elongation factor 1 delta
ankyrin repeat domain 19, pseudogene
Gene Aliases:
EF-1D, EF1D, FP1047
ANKRD19, bA526D8.2
Chromosome Location:
Chr.8: 143579697 - 143597675 on Build GRCh38
Chromosome Location:
Chr.9: 92809611 - 92838457 on Build GRCh38
UniGene ID:
Hs.333388
Hs.643597
Species:
Human
Species Specific ID (Flybase ID):
-
-
siRNA ID
HSS151388
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: EEF1D
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001130053.2
NP_001123525.2
9
2163
NM_001130055.2
NP_001123527.1
8
1234
NM_001130056.2
NP_001123528.1
6
986
NM_001130057.2
NP_001123529.1
7
1240
NM_001195203.1
NP_001182132.1
7
1001
NM_001289950.1
NP_001276879.1
7
1055
NM_001317743.1
NP_001304672.1
6
983
NM_001960.4
NP_001951.2
7
1058
NM_032378.4
NP_115754.3
9
2280
XM_005250820.3
XP_005250877.1
8
7155
XM_005250823.1
XP_005250880.1
8
1889
XM_006716519.3
XP_006716582.1
8
7152
XM_006716520.3
XP_006716583.1
7
7083
XM_006716522.1
XP_006716585.1
8
2143
XM_006716523.1
XP_006716586.1
9
2103
XM_006716524.1
XP_006716587.1
8
1910
XM_006716525.1
XP_006716588.1
7
1078
XM_011516905.2
XP_011515207.1
9
6725
XM_011516906.2
XP_011515208.1
11
5317
XM_017013168.1
XP_016868657.1
9
6156
XM_017013169.1
XP_016868658.1
10
5812
XM_017013170.1
XP_016868659.1
9
5598
XM_017013171.1
XP_016868660.1
8
1886
XM_017013172.1
XP_016868661.1
7
2071
XM_017013173.1
XP_016868662.1
6
1163
GenBank mRNA
AF370363.1
-
9
1863
AK024550.1
-
9
2017
AK093756.1
-
7
2200
AK225040.1
-
9
2018
AK225243.1
-
7
795
AK225270.1
-
7
796
AK293339.1
-
7
739
AK314311.1
-
9
1899
AL833379.1
-
3
3530
BC000678.2
-
6
1530
BC007847.2
-
9
1884
4129368
BC009907.2
-
1
780
BC012819.2
-
1
780
BC062535.1
-
7
772
BC071840.1
-
7
760
BC094806.1
-
6
697
BT007242.1
-
7
1821
Z21507.1
-
7
792
Gene Symbol: ANKRD19P
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NR_026868.1
9
1308
GenBank mRNA
AK093497.1
-
7
900
BC028712.1
-
9
1272
BC038951.1
-
8
1082
5733799
Back To Top
More Information
Panther Classification:
Molecular Function -
translation elongation factor
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
translational elongation
signal transduction
positive regulation of I-kappaB kinase/NF-kappaB signaling
cellular response to ionizing radiation
cell-cell adhesion
DNA binding
translation elongation factor activity
signal transducer activity
protein binding
translation factor activity, RNA binding
cadherin binding involved in cell-cell adhesion
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0