Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC2A14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286233.1 | NP_001273162.1 | 9 | 1296 | ||
NM_001286234.1 | NP_001273163.1 | 4 | 542 | |||
NM_001286235.1 | NP_001273164.1 | 6 | 840 | |||
NM_001286237.1 | NP_001273166.1 | 3 | 435 | |||
NM_153449.3 | NP_703150.1 | 5 | 578 | |||
XM_005253315.3 | XP_005253372.1 | 4 | 376 | |||
XM_005253317.4 | XP_005253374.1 | 4 | 895 | |||
XM_011520562.1 | XP_011518864.1 | 7 | 730 | |||
XM_011520563.2 | XP_011518865.1 | 4 | 390 | |||
XM_017018841.1 | XP_016874330.1 | 5 | 475 | |||
XM_017018842.1 | XP_016874331.1 | 8 | 1046 | |||
XM_017018843.1 | XP_016874332.1 | 8 | 1370 | |||
XM_017018844.1 | XP_016874333.1 | 5 | 425 | |||
XM_017018845.1 | XP_016874334.1 | 5 | 967 | |||
XM_017018846.1 | XP_016874335.1 | 5 | 551 | |||
XM_017018847.1 | XP_016874336.1 | 4 | 508 | |||
GenBank mRNA | AF481878.1 | - | 4 | 361 | ||
AF481879.1 | - | 5 | 438 | |||
AK122783.1 | - | 6 | 628 | |||
AK126026.1 | - | 9 | 1078 | |||
AK299037.1 | - | 3 | 387 | |||
AK316238.1 | - | 3 | 387 | |||
AL832448.1 | - | 2 | 202 | |||
AY357941.1 | - | 4 | 527 | |||
BC060766.1 | - | 4 | 389 | 5297510 | ||
BG717327.1 | - | 5 | 604 | |||
DA723309.1 | - | 3 | 435 | |||
DB082816.1 | - | 4 | 419 |