Search
Search

ISM2| Gene Symbol: ISM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182509.3 | NP_872315.2 | 6 | 1266 | ||
| NM_199296.2 | NP_954993.1 | 7 | 1612 | |||
| XM_011536489.1 | XP_011534791.1 | 7 | 1502 | |||
| GenBank mRNA | AJ583024.1 | - | 7 | 1573 | ||
| AK056709.1 | - | 8 | 1666 | |||
| AK075445.1 | - | 7 | 1595 | |||
| AK291600.1 | - | 7 | 1561 | |||
| AK300214.1 | - | 6 | 1318 | |||
| BC033140.1 | - | 6 | 1379 | |||
| BC101019.1 | - | 7 | 1561 | 40007143 | ||
| BC101020.1 | - | 7 | 1561 | 40007145 | ||
| BC101021.2 | - | 6 | 1215 | 40007146 | ||
| BC101022.2 | - | 7 | 1561 | 40007147 | ||
| CR749863.1 | - | 7 | 1561 | |||