Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OTOA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161683.1 | NP_001155155.1 | 12 | 1606 | ||
NM_144672.3 | NP_653273.3 | 16 | 1750 | |||
NM_170664.2 | NP_733764.1 | 7 | 844 | |||
XM_011545747.2 | XP_011544049.1 | 17 | 2129 | |||
XM_011545748.2 | XP_011544050.2 | 6 | 1123 | |||
XM_017022950.1 | XP_016878439.1 | 17 | 1853 | |||
XM_017022951.1 | XP_016878440.1 | 2 | 147 | |||
GenBank mRNA | AK093062.1 | - | 7 | 844 | ||
AK125840.1 | - | 12 | 1606 | |||
AK302733.1 | - | 6 | 906 | |||
BC129992.1 | - | 16 | 1747 | |||
BC129993.1 | - | 16 | 1754 | 40128985 |