Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DPY19L3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172774.1 | NP_001166245.1 | 2 | 241 | ||
NM_207325.2 | NP_997208.2 | 2 | 267 | |||
XM_011526526.2 | XP_011524828.1 | 2 | 222 | |||
XM_017026366.1 | XP_016881855.1 | 2 | 258 | |||
XM_017026367.1 | XP_016881856.1 | 2 | 225 | |||
GenBank mRNA | AF533710.1 | - | 2 | 194 | ||
AF533711.1 | - | 2 | 200 | |||
AK126757.1 | - | 2 | 258 | |||
AK129497.1 | - | 2 | 222 | |||
BC029162.1 | - | 2 | 200 | |||
BC131725.1 | - | 1 | 89 | 40123868 | ||
DA852420.1 | - | 2 | 241 | |||
DR005952.1 | - | 2 | 273 |