Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD18A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147195.2 | NP_671728.2 | 3 | 444 | ||
XM_006716749.3 | XP_006716812.1 | 3 | 667 | |||
XM_011517836.2 | XP_011516138.1 | 3 | 668 | |||
XM_011517837.2 | XP_011516139.1 | 3 | 668 | |||
XM_011517839.2 | XP_011516141.1 | 3 | 666 | |||
XM_017014576.1 | XP_016870065.1 | 3 | 667 | |||
XM_017014577.1 | XP_016870066.1 | 3 | 663 | |||
GenBank mRNA | AB095935.1 | - | 3 | 389 | ||
BC056266.1 | - | 3 | 745 | |||
BC152434.1 | - | 3 | 412 | 8862553 |