siRNA Details
Gene Symbol: FITM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_203402.2 | NP_981947.1 | 2 | 659 | ||
GenBank mRNA | BC139911.1 | - | 2 | 659 | 40147642 | |
BI114004.1 | - | 2 | 472 |

Gene Symbol: FITM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_203402.2 | NP_981947.1 | 2 | 659 | ||
GenBank mRNA | BC139911.1 | - | 2 | 659 | 40147642 | |
BI114004.1 | - | 2 | 472 |