Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133475.1 | NP_597732.1 | 9 | 864 | ||
XM_011527756.2 | XP_011526058.1 | 10 | 774 | |||
XM_011527757.2 | XP_011526059.1 | 9 | 1022 | |||
XM_011527758.2 | XP_011526060.1 | 9 | 687 | |||
XM_011527761.1 | XP_011526063.1 | 11 | 844 | |||
XM_011527764.2 | XP_011526066.1 | 8 | 935 | |||
XM_011527765.2 | XP_011526067.1 | 8 | 588 | |||
XM_017026438.1 | XP_016881927.1 | 11 | 835 | |||
XM_017026439.1 | XP_016881928.1 | 10 | 757 | |||
XM_017026440.1 | XP_016881929.1 | 9 | 730 | |||
GenBank mRNA | AB075861.1 | - | 7 | 561 | ||
BC028236.1 | - | 7 | 506 | |||
BI914435.1 | - | 7 | 558 |