Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PABPC1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001124756.2 | NP_001118228.1 | 6, 7 | 954 | ||
NR_134983.1 | 6, 7 | 954 | ||||
GenBank mRNA | BX451955.2 | - | 1, 2 | 115 | ||
EU190483.1 | - | 6, 7 | 890 |