Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RWDD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001307922.1 | NP_001294851.1 | 8 | 1247 | ||
NM_152682.3 | NP_689895.2 | 8 | 1251 | |||
GenBank mRNA | AK021702.1 | - | 8 | 1176 | ||
BC017472.1 | - | 8 | 1186 | 3909581 | ||
BC107432.1 | - | 8 | 1032 | 6104330 |