Search
Search

KDM1B| Gene Symbol: KDM1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153042.3 | NP_694587.3 | 7 | 743 | ||
| XM_005248925.1 | XP_005248982.1 | 7 | 743 | |||
| XM_005248926.1 | XP_005248983.1 | 7 | 743 | |||
| XM_005248928.1 | XP_005248985.1 | 7 | 743 | |||
| XM_011514385.1 | XP_011512687.1 | 7 | 743 | |||
| XM_011514386.1 | XP_011512688.1 | 7 | 743 | |||
| XM_017010440.1 | XP_016865929.1 | 7 | 743 | |||
| XM_017010441.1 | XP_016865930.1 | 7 | 743 | |||
| XM_017010442.1 | XP_016865931.1 | 6 | 589 | |||
| XM_017010443.1 | XP_016865932.1 | 6 | 536 | |||
| XM_017010445.1 | XP_016865934.1 | 7 | 743 | |||
| XM_017010446.1 | XP_016865935.1 | 7 | 743 | |||
| GenBank mRNA | AK125318.1 | - | 7 | 679 | ||