Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PCSK9 SIRNA Assays ›
          Gene Symbol:
          PCSK9
          Gene Name:
          proprotein convertase subtilisin/kexin type 9
          Gene Aliases:
          FH3, HCHOLA3, LDLCQ1, NARC-1, NARC1, PC9
          Chromosome Location:
          Chr.1: 55039476 - 55064853 on Build GRCh38
          UniGene ID:
          Hs.18844
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS153993
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PCSK9
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_174936.3 NP_777596.2 5, 6 1159
          NR_110451.1 3, 4 456
          GenBank mRNA AK075365.1 - 5, 6 1088
          AK122717.1 - 6, 7 844
          AK124635.1 - 2, 3 1618
          AK297473.1 - 3, 4 456
          EF692496.1 - 5, 6 797

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_199253, AY847775, BC133063.
          This siRNA also targets the following transcripts in Mouse: NM_153565, BC038085, AK043077, AK149520, AY273821.

          Panther Classification:

          Molecular Function -

          serine protease

          Gene Ontology Categories:

          Function(s) Process(es)

          kidney development
          liver development
          negative regulation of receptor recycling
          positive regulation of receptor internalization
          triglyceride metabolic process
          phospholipid metabolic process
          apoptotic process
          lysosomal transport
          cholesterol metabolic process
          cellular response to starvation
          regulation of receptor activity
          negative regulation of low-density lipoprotein particle clearance
          protein autoprocessing
          neurogenesis
          neuron differentiation
          low-density lipoprotein particle receptor catabolic process
          positive regulation of low-density lipoprotein particle receptor catabolic process
          cellular response to insulin stimulus
          lipoprotein metabolic process
          cholesterol homeostasis
          regulation of neuron apoptotic process
          positive regulation of neuron apoptotic process
          negative regulation of receptor activity
          negative regulation of sodium ion transmembrane transporter activity
          serine-type endopeptidase activity
          protein binding
          sodium channel inhibitor activity
          low-density lipoprotein particle binding
          receptor inhibitor activity
          apolipoprotein binding
          very-low-density lipoprotein particle binding
          apolipoprotein receptor binding
          protein self-association
          poly(A) RNA binding
          low-density lipoprotein particle receptor binding
          very-low-density lipoprotein particle receptor binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0