Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CRACR2B SIRNA Assays ›
          Gene Symbol:
          CRACR2B
          Gene Name:
          calcium release activated channel regulator 2B
          Gene Aliases:
          EFCAB4A
          Chromosome Location:
          Chr.11: 825558 - 831991 on Build GRCh38
          UniGene ID:
          Hs.660936
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS154237
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price Contact Us
          Your Price
          Online offer:
          3062.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CRACR2B
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001286606.1 NP_001273535.1 9 1578
          NM_173584.4 NP_775855.3 8 1411
          XM_011520011.1 XP_011518313.1 8 2351
          XM_011520019.1 XP_011518321.1 9 2160
          XM_017017581.1 XP_016873070.1 7 2634
          XM_017017582.1 XP_016873071.1 8 2441
          XM_017017583.1 XP_016873072.1 8 2413
          XM_017017584.1 XP_016873073.1 8 2453
          XM_017017585.1 XP_016873074.1 8 2992
          XM_017017586.1 XP_016873075.1 8 2313
          XM_017017587.1 XP_016873076.1 8 2684
          XM_017017588.1 XP_016873077.1 8 2108
          XM_017017589.1 XP_016873078.1 7 2616
          XM_017017590.1 XP_016873079.1 8 2223
          XM_017017591.1 XP_016873080.1 8 2193
          XM_017017592.1 XP_016873081.1 8 2772
          XM_017017593.1 XP_016873082.1 8 2093
          XM_017017594.1 XP_016873083.1 8 2464
          XM_017017595.1 XP_016873084.1 8 1888
          XM_017017596.1 XP_016873085.1 7 2396
          XM_017017597.1 XP_016873086.1 8 2443
          XM_017017598.1 XP_016873087.1 8 1993
          XM_017017599.1 XP_016873088.1 9 1650
          GenBank mRNA AK075180.1 - 5 1377
          AK097334.1 - 2 2194
          AK127926.1 - 5 1482
          BC033196.1 - 8 1192 4849453
          BC073821.1 - 9 1505 5952348
          BG674461.1 - 7 640
          BU621015.1 - 1 267
          BX104079.1 - 1 257

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          store-operated calcium entry
          cellular protein localization
          regulation of store-operated calcium entry
          calcium ion binding
          protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0