Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZIK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010879.3 | NP_001010879.2 | 3 | 534 | ||
NM_001321145.1 | NP_001308074.1 | 2 | 495 | |||
NM_001321146.1 | NP_001308075.1 | 2 | 718 | |||
XM_011526761.1 | XP_011525063.1 | 3 | 826 | |||
XM_011526762.1 | XP_011525064.1 | 3 | 1091 | |||
XM_011526763.1 | XP_011525065.1 | 2 | 787 | |||
XM_011526764.1 | XP_011525066.1 | 3 | 169 | |||
XM_011526766.2 | XP_011525068.1 | 2 | 108 | |||
GenBank mRNA | AK092538.1 | - | 3 | 368 | ||
AK128340.1 | - | 2 | 331 | |||
BC103958.1 | - | 3 | 250 | 40011910 | ||
BC103959.2 | - | 2 | 434 | 40011911 | ||
DB480028.2 | - | 3 | 486 |