Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC9C1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320531.1 | NP_001307460.1 | 7 | 915 | ||
NM_183061.2 | NP_898884.1 | 7 | 915 | |||
XM_006713603.2 | XP_006713666.1 | 7 | 950 | |||
XM_011512719.1 | XP_011511021.1 | 7 | 915 | |||
XM_011512720.1 | XP_011511022.1 | 7 | 915 | |||
XM_011512721.1 | XP_011511023.1 | 8 | 1093 | |||
XM_011512724.1 | XP_011511026.1 | 7 | 915 | |||
XM_011512726.2 | XP_011511028.1 | 7 | 915 | |||
XM_011512727.2 | XP_011511029.1 | 7 | 915 | |||
XM_017006246.1 | XP_016861735.1 | 5 | 618 | |||
XM_017006247.1 | XP_016861736.1 | 7 | 915 | |||
XM_017006248.1 | XP_016861737.1 | 7 | 915 | |||
XM_017006249.1 | XP_016861738.1 | 7 | 915 | |||
XM_017006250.1 | XP_016861739.1 | 7 | 915 | |||
GenBank mRNA | AK128084.1 | - | 7 | 915 | ||
BC044801.1 | - | 7 | 884 | 4827529 |