Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRID2IP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145118.1 | NP_001138590.1 | 20 | 3344 | ||
XM_011515404.2 | XP_011513706.1 | 19 | 2819 |