Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLTPD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014985.2 | NP_001014985.2 | 3 | 321 | ||
XM_005256626.3 | XP_005256683.1 | 2 | 472 | |||
GenBank mRNA | BC029290.1 | - | 3 | 321 | ||
BC150536.1 | - | 1 | 62 |
Gene Symbol: LOC284014 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK057919.1 | - | 1 | 784 | ||
AK095567.1 | - | 1 | 1214 |