Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIF1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015074.3 | NP_055889.2 | 47 | 7729 | ||
GenBank mRNA | AB017133.1 | - | 48 | 5905 | ||
AB088210.1 | - | 48 | 5821 | |||
AB088211.1 | - | 47 | 5779 | |||
AB088212.1 | - | 48 | 5743 | |||
AB088213.1 | - | 47 | 5701 | |||
AK022977.1 | - | 9 | 3278 | |||
AK025761.1 | - | 1 | 799 | |||
BC047632.1 | - | 1 | 125 | |||
DA113609.1 | - | 1 | 294 |