Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LDLRAD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010978.3 | NP_001010978.2 | 4 | 290 | ||
NM_001276392.1 | NP_001263321.1 | 2 | 109 | |||
NM_001276393.1 | NP_001263322.1 | 3 | 161 | |||
XM_011541446.1 | XP_011539748.1 | 4 | 270 | |||
GenBank mRNA | BC127708.1 | - | 4 | 287 | 40127476 | |
BC127709.1 | - | 2 | 106 | 40127477 | ||
BC140760.1 | - | 3 | 105 | 9021628 | ||
BC144466.1 | - | 3 | 105 | 9052992 | ||
CF887342.1 | - | 4 | 290 |