Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC6A17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010898.2 | NP_001010898.1 | 11, 12 | 2284 | ||
XM_006710643.2 | XP_006710706.1 | 11, 12 | 1992 | |||
GenBank mRNA | AK289982.1 | - | 11, 12 | 2280 | ||
AK297429.1 | - | 2, 3 | 267 | |||
BC140908.1 | - | 10, 11 | 1869 | 9056720 |