Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MROH2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287395.1 | NP_001274324.1 | 15 | 1790 | ||
XM_011511075.2 | XP_011509377.1 | 15 | 1967 | |||
XM_011511076.2 | XP_011509378.1 | 15 | 1967 | |||
XM_011511077.2 | XP_011509379.1 | 15 | 1967 | |||
XM_011511078.2 | XP_011509380.1 | 15 | 1967 | |||
XM_011511079.2 | XP_011509381.1 | 15 | 1967 | |||
XM_011511080.2 | XP_011509382.1 | 15 | 3438 | |||
XM_011511081.2 | XP_011509383.1 | 15 | 1967 | |||
XM_011511082.2 | XP_011509384.1 | 15 | 1967 | |||
XM_011511083.2 | XP_011509385.1 | 15 | 1967 | |||
XM_011511084.2 | XP_011509386.1 | 15 | 1967 | |||
XM_011511085.2 | XP_011509387.1 | 9 | 1037 | |||
XM_011511086.2 | XP_011509388.1 | 15 | 1967 |