Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RUBCN SIRNA Assays ›
          Gene Symbol:
          RUBCN CNOT10 DNAJC5G FAM53C FUT2 LINC00861 LOC284240 LRP2BP MEG3 MPC1 ORC4
          Gene Name:
          RUN and cysteine rich domain containing beclin 1 interacting protein
          CCR4-NOT transcription complex subunit 10
          DnaJ heat shock protein family (Hsp40) member C5 gamma
          family with sequence similarity 53 member C
          fucosyltransferase 2
          long intergenic non-protein coding RNA 861
          uncharacterized LOC284240
          LRP2 binding protein
          maternally expressed 3 (non-protein coding)
          mitochondrial pyruvate carrier 1
          origin recognition complex subunit 4
          Gene Aliases:
          KIAA0226, RUBICON, SCAR15
          -
          CSP-gamma
          C5orf6
          B12QTL1, SE, SEC2, Se2, sej
          -
          -
          -
          FP504, GTL2, LINC00023, NCRNA00023, PRO0518, PRO2160, onco-lncRNA-83, prebp1
          BRP44L, CGI-129, MPYCD, dJ68L15.3
          ORC4L, ORC4P
          Chromosome Location:
          Chr.3: 197670213 - 197749851 on Build GRCh38
          Chromosome Location:
          Chr.3: 32685145 - 32773875 on Build GRCh38
          Chromosome Location:
          Chr.2: 27275421 - 27281430 on Build GRCh38
          Chromosome Location:
          Chr.5: 138337535 - 138349729 on Build GRCh38
          Chromosome Location:
          Chr.19: 48695971 - 48705934 on Build GRCh38
          Chromosome Location:
          Chr.8: 125922523 - 125951197 on Build GRCh38
          Chromosome Location:
          Chr.18: 79573705 - 79591904 on Build GRCh38
          Chromosome Location:
          Chr.4: 185363878 - 185397160 on Build GRCh38
          Chromosome Location:
          Chr.14: 100826108 - 100861026 on Build GRCh38
          Chromosome Location:
          Chr.6: 166364919 - 166383013 on Build GRCh38
          Chromosome Location:
          Chr.2: 147930397 - 148021604 on Build GRCh38
          UniGene ID:
          Hs.478868
          Hs.444851
          Hs.116303
          Hs.54056
          Hs.579928
          Hs.729642
          Hs.448912
          Hs.558513
          Hs.654863
          Hs.172755
          Hs.558364
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          siRNA ID HSS158512
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: RUBCN
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001145642.3 NP_001139114.1 21 5423
          NM_014687.2 NP_055502.1 20 5219
          XM_005269374.3 XP_005269431.1 21 5259
          XM_006713827.3 XP_006713890.1 22 5335
          XM_006713828.3 XP_006713891.1 21 5289
          XM_006713829.3 XP_006713892.1 22 5650
          XM_006713830.3 XP_006713893.1 21 5183
          XM_006713831.3 XP_006713894.1 21 5496
          XM_017007543.1 XP_016863032.1 20 5134
          XM_017007544.1 XP_016863033.1 20 5105
          XM_017007545.1 XP_016863034.1 20 5421
          GenBank mRNA BU501990.1 - 1 750
          D86979.3 - 22 4456
          Gene Symbol: CNOT10
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_046352.1 16 2132
          GenBank mRNA AK309670.1 - 16 2057
          Gene Symbol: DNAJC5G
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA AK311597.1 - 2 217
          Gene Symbol: FAM53C
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA AK026729.1 - 2 1670
          Gene Symbol: FUT2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000511.5 NP_000502.4 2 2504
          NM_001097638.2 NP_001091107.1 2 2494
          GenBank mRNA BC121066.1 - 3 1318
          D87942.1 - 2 2477
          Gene Symbol: LINC00861
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_038446.1 2 1913
          GenBank mRNA BX648371.1 - 2 1905
          Gene Symbol: LOC284240
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA BC017478.1 - 1 3116
          Gene Symbol: LRP2BP
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA BC150161.1 - 8 2959 40126591
          Gene Symbol: MEG3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_046473.1 7 5753
          Gene Symbol: MPC1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_011535894.1 XP_011534196.1 1 549
          XM_011535895.1 XP_011534197.1 1 549
          GenBank mRNA AK310934.1 - 2 1166
          Gene Symbol: ORC4
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA AK055257.1 - 1 556

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          glycosyltransferase

          Gene Ontology Categories:

          Function(s) Process(es)

          immune system process
          phagocytosis
          negative regulation of autophagy
          negative regulation of phosphatidylinositol 3-kinase activity
          negative regulation of endocytosis
          multivesicular body sorting pathway
          autophagosome maturation
          negative regulation of autophagosome maturation
          nuclear-transcribed mRNA poly(A) tail shortening
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          regulation of translation
          DNA damage response, signal transduction by p53 class mediator resulting in cell cycle arrest
          gene silencing by RNA
          carbohydrate metabolic process
          protein glycosylation
          fucosylation
          L-fucose catabolic process
          biological_process
          mitochondrial acetyl-CoA biosynthetic process from pyruvate
          mitochondrial pyruvate transmembrane transport
          protein binding
          galactoside 2-alpha-L-fucosyltransferase activity
          fucosyltransferase activity
          molecular_function
          pyruvate transmembrane transporter activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0