Search
Search

LOC101928195
LOC100996643
LOC101929583
LOC102724904
LOC105379443| Gene Symbol: LOC101928195 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135597.1 | 2 | 224 | |||
| GenBank mRNA | CR740458.1 | - | 2 | 224 | ||
| Gene Symbol: LOC100996643 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006716912.3 | XP_006716975.1 | 2 | 153 | ||
| Gene Symbol: LOC101929583 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_136299.1 | 1 | 16 | |||
| GenBank mRNA | DN831597.1 | - | 1 | 16 | ||
| Gene Symbol: LOC102724904 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011518105.2 | XP_011516407.1 | 1 | 119 | ||
| Gene Symbol: LOC105379443 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017015396.1 | XP_016870885.1 | 1 | 170 | ||