Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TUBA1C SIRNA Assays ›
          Gene Symbol:
          TUBA1C TUBA1A TUBA1B TUBA4B
          Gene Name:
          tubulin alpha 1c
          tubulin alpha 1a
          tubulin alpha 1b
          tubulin alpha 4b
          Gene Aliases:
          TUBA6, bcm948
          B-ALPHA-1, LIS3, TUBA3
          K-ALPHA-1
          TUBA4
          Chromosome Location:
          Chr.12: 49227926 - 49273338 on Build GRCh38
          Chromosome Location:
          Chr.12: 49184795 - 49189324 on Build GRCh38
          Chromosome Location:
          Chr.12: 49127782 - 49131521 on Build GRCh38
          Chromosome Location:
          Chr.2: 219253243 - 219272188 on Build GRCh38
          UniGene ID:
          Hs.652390
          Hs.654422
          Hs.524390
          Hs.664469
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          siRNA ID HSS159067
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price Contact Us
          Your Price
          Online offer:
          3062.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: TUBA1C
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001303114.1 NP_001290043.1 3 525
          NM_001303115.1 NP_001290044.1 5 758
          NM_001303116.1 NP_001290045.1 4 640
          NM_001303117.1 NP_001290046.1 4 643
          NM_032704.4 NP_116093.1 3 470
          GenBank mRNA AK223024.1 - 3 387
          AK293589.1 - 3 525
          AL539200.3 - 4 636
          BC004949.1 - 3 367 3617714
          BC005946.1 - 3 387 4134187
          BC011790.1 - 3 368 4025017
          BC019298.2 - 3 349 2905506
          BC021088.1 - 3 348 3543481
          BC033064.1 - 3 363 3926854
          BC051297.1 - 3 363 3832363
          BC063036.1 - 3 409 4397636
          BG469587.1 - 3 562
          DC366449.1 - 4 526
          Gene Symbol: TUBA1A
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA AK289483.1 - 3 559
          BC050637.2 - 3 371 6050536
          Gene Symbol: TUBA1B
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_006082.2 NP_006073.2 3 508
          GenBank mRNA AF081484.1 - 2 287
          AK026516.1 - 3 382
          AK026632.1 - 3 382
          AK056693.1 - 2 1290
          AK057773.1 - 3 382
          AK094717.1 - 2 1955
          AK098357.1 - 3 382
          AK098635.1 - 3 382
          AK098771.1 - 3 382
          AK289354.1 - 3 382
          BC000696.2 - 3 373 3349131
          BC001128.1 - 3 366 2964447
          BC006379.2 - 3 368 4100566
          BC006481.1 - 3 368 2905305
          BC008659.2 - 3 345 3871729
          BC009314.2 - 3 367 4111591
          BC009509.2 - 3 369 3533174
          BC009512.2 - 3 366 3537301
          BC009513.2 - 3 370 3537827
          BC010494.1 - 3 368 3445903
          BC011572.1 - 3 368 4561240
          BC015883.2 - 3 326 3909374
          BC017004.1 - 3 338 4430457
          BC018948.2 - 3 368
          BC021564.1 - 3 368 4579604
          BC030820.1 - 3 379 4277263
          BC071904.1 - 3 373 6089096
          BF979331.1 - 519
          DQ400107.1 - 3 372
          K00558.1 - 3 354
          S62639.1 - 2 262
          Gene Symbol: TUBA4B
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_003063.1 3 269
          GenBank mRNA AK024002.1 - 3 269

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_001044270, NM_022298, NM_001011995, XM_003749354, FQ210110, and others.

          Panther Classification:

          Molecular Function -

          tubulin

          Gene Ontology Categories:

          Function(s) Process(es)

          cytoskeleton organization
          microtubule-based process
          cytoskeleton-dependent intracellular transport
          cell division
          microtubule cytoskeleton organization
          cellular response to interleukin-4
          biological_process
          GTPase activity
          structural molecule activity
          structural constituent of cytoskeleton
          protein binding
          GTP binding
          double-stranded RNA binding
          ubiquitin protein ligase binding
          molecular_function

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0