Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPRNP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033789.2 | 2 | 1166 | |||
GenBank mRNA | AK126615.1 | - | 2 | 1166 | ||
BC136964.1 | - | 2 | 1043 | 9020956 |