Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ?
Create Account
Applications
Real-Time PCR
Oligos, Primers, Probes and Genes
Cloning
Protein Biology
Cell Culture and Transfection
Cell and Gene Therapy
Cell Analysis
Mass Spectrometry
Chromatography
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Online Order
Custom Primers & TaqMan Probes
miRNA Mimics & Inhibitors
Stealth RNAi / siRNA
Silencer Select siRNAs
Custom DNA Oligos
GeneArt Services
GeneArt Strings DNA Fragments
TrueGuide CRISPR gRNA
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all online orderable products
Services
Custom Services
Instrument Qualification Services
Technical Services
Enterprise Services
Pipette Services
Sample Request
Instrument Maintenance Services
Repairs and Relocation Services
OEM and Licensing Services
Events, Seminars, Training
See all services
Support
Catalogs
Application Notes
Safety Data Sheets
Legal and Regulatory
Certificates of Analysis Search
Nunc/Nalgene Product Certificates
e-learning
FAQs
Learning Centers
Technical Support Centers
See all help and support topics
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign In
Sign In
Don't have an account ?
Create Account
Connect Your Lab
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ANKRD36B SIRNA Assays ›
Gene Symbol:
ANKRD36B
ANKRD36
Gene Name:
ankyrin repeat domain 36B
ankyrin repeat domain 36
Gene Aliases:
KIAA1641
UNQ2430
Chromosome Location:
Chr.2: 97492663 - 97590245 on Build GRCh38
Chromosome Location:
Chr.2: 97113186 - 97264521 on Build GRCh38
UniGene ID:
Hs.532921
Hs.541894
Species:
Human
Species Specific ID (Flybase ID):
-
-
siRNA ID
HSS160246
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Contact Us
Your Price
Online offer:
3062.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ANKRD36B
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_025190.3
NP_079466.3
42
4277
XM_005263995.4
XP_005264052.1
42
4557
XM_006712661.1
XP_006712724.1
42
4557
XM_017004578.1
XP_016860067.1
42
4554
XM_017004579.1
XP_016860068.1
40
4455
XM_017004580.1
XP_016860069.1
40
4455
XM_017004581.1
XP_016860070.1
40
4455
XM_017004582.1
XP_016860071.1
40
4455
XM_017004583.1
XP_016860072.1
41
4419
XM_017004584.1
XP_016860073.1
36
4251
XM_017004585.1
XP_016860074.1
40
4312
XM_017004586.1
XP_016860075.1
37
3298
XM_017004587.1
XP_016860076.1
41
4486
XM_017004588.1
XP_016860077.1
40
4351
XM_017004589.1
XP_016860078.1
39
4244
XM_017004590.1
XP_016860079.1
38
4171
XM_017004591.1
XP_016860080.1
24
4275
XM_017004592.1
XP_016860081.1
43
4601
XM_017004593.1
XP_016860082.1
41
4407
XM_017004594.1
XP_016860083.1
40
4300
XM_017004595.1
XP_016860084.1
41
4356
GenBank mRNA
AF432209.1
-
3
1081
AK024934.1
-
4
389
AK056042.1
-
3
435
BC125132.1
-
4
389
BC125133.1
-
4
389
DB105321.1
-
4
436
Gene Symbol: ANKRD36
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001164315.1
NP_001157787.1
74
6005
XM_006712514.3
XP_006712577.1
74
6237
XM_011511130.1
XP_011509432.1
74
6237
XM_017004009.1
XP_016859498.1
74
6237
XM_017004010.1
XP_016859499.1
74
6234
XM_017004011.1
XP_016859500.1
72
6135
XM_017004012.1
XP_016859501.1
72
6135
XM_017004013.1
XP_016859502.1
72
6135
XM_017004014.1
XP_016859503.1
72
6135
XM_017004015.1
XP_016859504.1
72
6135
XM_017004016.1
XP_016859505.1
72
6135
XM_017004017.1
XP_016859506.1
72
6135
XM_017004018.1
XP_016859507.1
72
6135
XM_017004019.1
XP_016859508.1
72
6135
XM_017004020.1
XP_016859509.1
72
6135
XM_017004021.1
XP_016859510.1
72
6135
XM_017004022.1
XP_016859511.1
72
6135
XM_017004023.1
XP_016859512.1
72
6135
XM_017004024.1
XP_016859513.1
73
6099
XM_017004025.1
XP_016859514.1
70
6033
XM_017004026.1
XP_016859515.1
70
6033
XM_017004027.1
XP_016859516.1
70
6033
XM_017004028.1
XP_016859517.1
70
6033
XM_017004029.1
XP_016859518.1
70
6033
XM_017004030.1
XP_016859519.1
70
6033
XM_017004031.1
XP_016859520.1
70
6033
XM_017004032.1
XP_016859521.1
70
6033
XM_017004033.1
XP_016859522.1
70
6033
XM_017004034.1
XP_016859523.1
70
6033
XM_017004035.1
XP_016859524.1
70
6033
XM_017004036.1
XP_016859525.1
70
6033
XM_017004037.1
XP_016859526.1
70
6033
XM_017004038.1
XP_016859527.1
70
6033
XM_017004039.1
XP_016859528.1
68
5931
XM_017004040.1
XP_016859529.1
68
5931
XM_017004041.1
XP_016859530.1
68
5931
XM_017004042.1
XP_016859531.1
68
5931
XM_017004043.1
XP_016859532.1
68
5931
XM_017004044.1
XP_016859533.1
66
5829
XM_017004045.1
XP_016859534.1
66
5829
XM_017004046.1
XP_016859535.1
72
5992
XM_017004047.1
XP_016859536.1
71
5924
XM_017004048.1
XP_016859537.1
70
5851
XM_017004049.1
XP_016859538.1
72
6026
XM_017004050.1
XP_016859539.1
57
4430
XM_017004051.1
XP_016859540.1
55
4644
Back To Top
More Information
Gene Ontology Categories:
Process(es)
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0