Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRCD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017025013.1 | XP_016880502.1 | 1 | 471 | ||
XM_017025014.1 | XP_016880503.1 | 1 | 471 | |||
XM_017025015.1 | XP_016880504.1 | 1 | 471 |