Search
Search

| Gene Symbol: SLC35E2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182838.2 | NP_878258.1 | 6 | 3372 | ||
| GenBank mRNA | AK092315.1 | - | 1 | 920 | ||
| AK126501.1 | - | 1 | 1299 | |||
| Gene Symbol: SLC35E2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001110781.2 | NP_001104251.1 | 9 | 3102 | ||
| NM_001290264.1 | NP_001277193.1 | 10 | 3520 | |||
| XM_011542072.1 | XP_011540374.1 | 5 | 2463 | |||
| GenBank mRNA | AB007916.3 | - | 9 | 3040 | ||
| AK092311.1 | - | 3 | 2241 | |||
| AK125619.1 | - | 9 | 3026 | |||
| BC110653.1 | - | 9 | 3078 | 4392812 | ||
| CR936618.1 | - | 10 | 3239 | |||