Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EML6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039753.2 | NP_001034842.2 | 41 | 5980 | ||
XM_017004098.1 | XP_016859587.1 | 43 | 6157 | |||
XM_017004099.1 | XP_016859588.1 | 42 | 6142 | |||
XM_017004101.1 | XP_016859590.1 | 37 | 4991 | |||
GenBank mRNA | AK026400.1 | - | 1 | 550 | ||
AK124553.1 | - | 18 | 2010 | |||
AK131467.1 | - | 12 | 1853 |