Search
Search

| Gene Symbol: CREB5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011515618.2 | XP_011513920.1 | 2 | 686 | ||
| XM_017012807.1 | XP_016868296.1 | 2 | 759 | |||
| Gene Symbol: LOC401317 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC087859.1 | - | 2 | 674 | ||