Search
Search

ORAI2| Gene Symbol: ORAI2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001126340.2 | NP_001119812.1 | 4 | 4247 | ||
| NM_001271818.1 | NP_001258747.1 | 4 | 4270 | |||
| NM_001271819.1 | NP_001258748.1 | 3 | 4009 | |||
| NM_032831.3 | NP_116220.1 | 3 | 4138 | |||