Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ACOT11 SIRNA Assays ›
          Gene Symbol:
          ACOT11 ANKRD26 ATP6AP1 ATP8B3 CEP89 DPY19L3 ERAP2 GGT1 KAT5 LINC01242 LOC100289518 LOC101928058 LOC727721 NUGGC PPARA TJP2
          Gene Name:
          acyl-CoA thioesterase 11
          ankyrin repeat domain 26
          ATPase H+ transporting accessory protein 1
          ATPase phospholipid transporting 8B3
          centrosomal protein 89
          dpy-19 like 3 (C. elegans)
          endoplasmic reticulum aminopeptidase 2
          gamma-glutamyltransferase 1
          lysine acetyltransferase 5
          long intergenic non-protein coding RNA 1242
          immunoglobin superfamily member 21 pseudogene
          uncharacterized LOC101928058
          uncharacterized LOC727721
          nuclear GTPase, germinal center associated
          peroxisome proliferator activated receptor alpha
          tight junction protein 2
          Gene Aliases:
          BFIT, STARD14, THEA, THEM1
          THC2, bA145E8.1
          16A, ATP6IP1, ATP6S1, Ac45, CF2, VATPS1, XAP-3, XAP3
          ATPIK
          CCDC123, CEP123
          -
          L-RAP, LRAP
          CD224, D22S672, D22S732, GGT, GGT 1, GTG
          ESA1, HTATIP, HTATIP1, PLIP, TIP, TIP60, ZC2HC5, cPLA2
          -
          -
          -
          -
          C8orf80, HMFN0672, SLIP-GC
          NR1C1, PPAR, PPARalpha, hPPAR
          C9DUPq21.11, DFNA51, DUP9q21.11, PFIC4, X104, ZO2
          Chromosome Location:
          Chr.1: 54548134 - 54634744 on Build GRCh38
          Chromosome Location:
          Chr.10: 26938195 - 27100498 on Build GRCh38
          Chromosome Location:
          Chr.X: 154428632 - 154436517 on Build GRCh38
          Chromosome Location:
          Chr.19: 1782070 - 1812276 on Build GRCh38
          Chromosome Location:
          Chr.19: 32875925 - 32972029 on Build GRCh38
          Chromosome Location:
          Chr.19: 32405749 - 32485893 on Build GRCh38
          Chromosome Location:
          Chr.5: 96875940 - 96919703 on Build GRCh38
          Chromosome Location:
          Chr.22: 24583750 - 24629005 on Build GRCh38
          Chromosome Location:
          Chr.11: 65711996 - 65719606 on Build GRCh38
          Chromosome Location:
          Chr.9: 30388935 - 30408454 on Build GRCh38
          Chromosome Location:
          Chr.11: 86283927 - 86284671 on Build GRCh38
          Chromosome Location:
          Chr.8: 1971179 - 1973345 on Build GRCh38
          Chromosome Location:
          -
          Chromosome Location:
          Chr.8: 28021964 - 28084068 on Build GRCh38
          Chromosome Location:
          Chr.22: 46150534 - 46243756 on Build GRCh38
          Chromosome Location:
          Chr.9: 69099907 - 69255208 on Build GRCh38
          UniGene ID:
          Hs.745173
          Hs.361041
          Hs.633817
          Hs.306212
          Hs.599703
          Hs.194392
          Hs.482910
          Hs.595809
          Hs.397010
          Hs.680131
          Hs.528103
          -
          Hs.699065
          Hs.370129
          Hs.103110
          Hs.50382
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          siRNA ID HSS163289
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ACOT11
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA BC001517.2 - 8 2396
          Gene Symbol: ANKRD26
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_017015930.1 XP_016871419.1 37 8290
          Gene Symbol: ATP6AP1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA AK090462.1 - 2 1265
          Gene Symbol: ATP8B3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA AK125225.1 - 14 2408
          Gene Symbol: CEP89
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_005259344.3 XP_005259401.1 19 3380
          XM_011527425.2 XP_011525727.1 16 2904
          Gene Symbol: DPY19L3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA BC029162.1 - 3 949
          Gene Symbol: ERAP2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_011543544.1 XP_011541846.1 18 3261
          GenBank mRNA AB163917.1 - 19 3398
          AK074213.1 - 4 690
          AY028805.1 - 18 3283
          BC065240.1 - 18 3156 6166289
          Gene Symbol: GGT1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA AL832738.1 - 5267
          Gene Symbol: KAT5
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA AK130717.1 - 1 1988
          Gene Symbol: LINC01242
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA BC022036.1 - 2 659
          Gene Symbol: LOC100289518
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA AK026856.1 - 2 1863
          Gene Symbol: LOC101928058
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_136274.1 3 352
          NR_136275.1 2 230
          GenBank mRNA DA540491.1 - 2 500
          Gene Symbol: LOC727721
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA DA084091.1 - 457
          Gene Symbol: NUGGC
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001010906.1 NP_001010906.1 19 2814
          XM_011544523.2 XP_011542825.1 19 3083
          XM_011544524.2 XP_011542826.1 19 4589
          XM_011544525.1 XP_011542827.1 11 1645
          GenBank mRNA AK129923.1 - 5 915
          Gene Symbol: PPARA
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA BC000052.2 - 6 1272
          Gene Symbol: TJP2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA AK304447.1 - 3 213

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          esterase ATP synthase primary active transporter metalloprotease

          Gene Ontology Categories:

          Function(s) Process(es)

          mitochondrion organization
          chemical synaptic transmission
          nonmotile primary cilium assembly
          cilium assembly
          adaptive immune response
          antigen processing and presentation of peptide antigen via MHC class I
          proteolysis
          regulation of blood pressure
          antigen processing and presentation of endogenous peptide antigen via MHC class I
          peptide catabolic process
          cellular response to DNA damage stimulus
          somatic hypermutation of immunoglobulin genes
          protein binding
          endopeptidase activity
          aminopeptidase activity
          metallopeptidase activity
          zinc ion binding
          peptide binding
          metalloaminopeptidase activity
          GTP binding
          hydrolase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-4x8c5:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0