Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC105369140 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017003053.1 | XP_016858542.1 | 6 | 1732 |
Gene Symbol: LINC00869 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK090660.1 | - | 5 | 1204 |