Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other NUDT4P1 SIRNA Assays ›
          Gene Symbol:
          NUDT4P1 FAM188A LINC00578 LOC645645 MKRN3 NUDT4 NUDT4P2 PTAFR SLC25A51 STK11
          Gene Name:
          nudix hydrolase 4 pseudogene 1
          family with sequence similarity 188 member A
          long intergenic non-protein coding RNA 578
          uncharacterized LOC645645
          makorin ring finger protein 3
          nudix hydrolase 4
          nudix hydrolase 4 pseudogene 2
          platelet activating factor receptor
          solute carrier family 25 member 51
          serine/threonine kinase 11
          Gene Aliases:
          -
          C10orf97, CARP, DERP5, MST126, MSTP126, my042
          -
          -
          CPPB2, D15S9, RNF63, ZFP127, ZNF127
          DIPP2, DIPP2alpha, DIPP2beta, HDCMB47P
          -
          PAFR
          CG7943, MCART1
          LKB1, PJS, hLKB1
          Chromosome Location:
          Chr.1: 148748600 - 148752401 on Build GRCh38
          Chromosome Location:
          Chr.10: 15778169 - 15860533 on Build GRCh38
          Chromosome Location:
          Chr.3: 177441921 - 177752704 on Build GRCh38
          Chromosome Location:
          -
          Chromosome Location:
          Chr.15: 23565307 - 23568020 on Build GRCh38
          Chromosome Location:
          Chr.12: 93377923 - 93403248 on Build GRCh38
          Chromosome Location:
          Chr.1: 146297773 - 146301610 on Build GRCh38
          Chromosome Location:
          Chr.1: 28147166 - 28193936 on Build GRCh38
          Chromosome Location:
          Chr.9: 37877575 - 37904353 on Build GRCh38
          Chromosome Location:
          Chr.19: 1205799 - 1228435 on Build GRCh38
          UniGene ID:
          Hs.601274
          Hs.158870
          Hs.581170
          Hs.159711
          Hs.679587
          Hs.506325
          Hs.601274
          Hs.77542
          Hs.634120
          Hs.515005
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          siRNA ID HSS163572
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: NUDT4P1
          Interrogated Sequence Translated Protein RNADB ID Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_002212.4
          1 2345
          GenBank mRNA AF311215.1 - LIT2089
          NUDT4 pseudogene
          5 2267
          Gene Symbol: FAM188A
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_011519691.2 XP_011517993.1 14 3240
          Gene Symbol: LINC00578
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_047568.1 4 965
          Gene Symbol: LOC645645
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA AK126090.1 - 1 2113
          Gene Symbol: MKRN3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA BC070293.1 - 2 1109
          Gene Symbol: NUDT4
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001301022.1 NP_001287951.1 5 2133
          NM_001301023.1 NP_001287952.1 5 2130
          NM_001301024.1 NP_001287953.1 5 2033
          NM_019094.5 NP_061967.3 5 2388
          NM_199040.3 NP_950241.1 5 2391
          XM_011537794.2 XP_011536096.1 5 1931
          XM_011537795.2 XP_011536097.1 6 3943
          XM_011537796.2 XP_011536098.1 6 2072
          GenBank mRNA AB007956.2 - 6 2187
          AF191651.1 - 5 1981
          AF191652.2 - 5 1979
          AF191654.2 - 1 67
          BC012069.1 - 5 2180 4650281
          CR749445.1 - 5 2103
          Gene Symbol: NUDT4P2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_104005.1 1 2389
          GenBank mRNA AK074343.1 - 5 2391
          Gene Symbol: PTAFR
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000952.4 NP_000943.1 2 2730
          NM_001164721.1 NP_001158193.1 3 2674
          NM_001164722.2 NP_001158194.1 2 2592
          NM_001164723.2 NP_001158195.1 3 2801
          Gene Symbol: SLC25A51
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_024872.2 5 745
          NR_024873.2 4 718
          GenBank mRNA AK091407.1 - 5 494
          AK098185.1 - 4 718
          Gene Symbol: STK11
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA AK128518.1 - 1 1388

          Back To Top

          More Information


          Set Membership:

          G-protein coupled receptor

          Panther Classification:

          Molecular Function -

          ubiquitin-protein ligase phosphatase G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          apoptotic process
          cyclic nucleotide metabolic process
          calcium-mediated signaling
          cyclic-nucleotide-mediated signaling
          intracellular signal transduction
          inositol phosphate metabolic process
          regulation of RNA export from nucleus
          intracellular transport
          cytokine production
          regulation of transcription from RNA polymerase II promoter
          chemotaxis
          inflammatory response
          immune response
          G-protein coupled receptor signaling pathway
          parturition
          response to symbiotic bacterium
          positive regulation of phospholipase C activity
          lipopolysaccharide-mediated signaling pathway
          positive regulation of tumor necrosis factor production
          inositol trisphosphate biosynthetic process
          G-protein coupled purinergic nucleotide receptor signaling pathway
          positive regulation of neutrophil degranulation
          transcytosis
          positive regulation of interleukin-6 biosynthetic process
          positive regulation of translation
          negative regulation of blood pressure
          positive regulation of vasoconstriction
          positive regulation of vasodilation
          phosphatidylinositol-mediated signaling
          positive regulation of smooth muscle cell proliferation
          interferon-gamma-mediated signaling pathway
          positive regulation of inositol phosphate biosynthetic process
          cellular response to gravity
          cellular response to cAMP
          cellular response to fatty acid
          response to dexamethasone
          positive regulation of voltage-gated chloride channel activity
          positive regulation of leukocyte tethering or rolling
          positive regulation of sensory perception of pain
          positive regulation of transcytosis
          positive regulation of maternal process involved in parturition
          positive regulation of gastro-intestinal system smooth muscle contraction
          cellular response to 2-O-acetyl-1-O-hexadecyl-sn-glycero-3-phosphocholine
          translation
          transport
          protein binding
          diphosphoinositol-polyphosphate diphosphatase activity
          snoRNA binding
          metal ion binding
          m7G(5')pppN diphosphatase activity
          inositol diphosphate tetrakisphosphate diphosphatase activity
          inositol bisdiphosphate tetrakisphosphate diphosphatase activity
          inositol diphosphate pentakisphosphate diphosphatase activity
          inositol-1-diphosphate-2,3,4,5,6-pentakisphosphate diphosphatase activity
          inositol-3-diphosphate-1,2,4,5,6-pentakisphosphate diphosphatase activity
          inositol-5-diphosphate-1,2,3,4,6-pentakisphosphate diphosphatase activity
          inositol-1,5-bisdiphosphate-2,3,4,6-tetrakisphosphate 1-diphosphatase activity
          inositol-1,5-bisdiphosphate-2,3,4,6-tetrakisphosphate 5-diphosphatase activity
          inositol-3,5-bisdiphosphate-2,3,4,6-tetrakisphosphate 5-diphosphatase activity
          lipopolysaccharide binding
          lipopolysaccharide receptor activity
          G-protein coupled receptor activity
          platelet activating factor receptor activity
          phospholipid binding
          G-protein coupled purinergic nucleotide receptor activity
          mitogen-activated protein kinase binding
          structural constituent of ribosome

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0