Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGPD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024457.3 | NP_001019628.3 | 23 | 6113 | ||
GenBank mRNA | CR749330.1 | - | 23 | 6113 |