Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CYB5D2 SIRNA Assays ›
          Gene Symbol:
          CYB5D2 DBF4B IAPP KIF1B LAMC3 LINC00269 PCYT1A RBM23 SP110 ZNHIT1
          Gene Name:
          cytochrome b5 domain containing 2
          DBF4 zinc finger B
          islet amyloid polypeptide
          kinesin family member 1B
          laminin subunit gamma 3
          long intergenic non-protein coding RNA 269
          phosphate cytidylyltransferase 1, choline, alpha
          RNA binding motif protein 23
          SP110 nuclear body protein
          zinc finger HIT-type containing 1
          Gene Aliases:
          -
          ASKL1, CHIFB, DRF1, ZDBF1B
          DAP, IAP
          CMT2, CMT2A, CMT2A1, HMSNII, KLP, NBLST1
          OCCM
          CXorf62, NCRNA00269
          CCTA, CT, CTA, CTPCT, PCYT1, SMDCRD
          CAPERbeta, PP239, RNPC4
          IFI41, IFI75, IPR1, VODI
          CG1I, ZNFN4A1
          Chromosome Location:
          Chr.17: 4143117 - 4157701 on Build GRCh38
          Chromosome Location:
          Chr.17: 44708581 - 44752732 on Build GRCh38
          Chromosome Location:
          Chr.12: 21354928 - 21379980 on Build GRCh38
          Chromosome Location:
          Chr.1: 10210706 - 10381603 on Build GRCh38
          Chromosome Location:
          Chr.9: 131009117 - 131093059 on Build GRCh38
          Chromosome Location:
          Chr.X: 69179557 - 69209924 on Build GRCh38
          Chromosome Location:
          Chr.3: 196237745 - 196287752 on Build GRCh38
          Chromosome Location:
          Chr.14: 22900645 - 22919187 on Build GRCh38
          Chromosome Location:
          Chr.2: 230165466 - 230225729 on Build GRCh38
          Chromosome Location:
          Chr.7: 101217704 - 101224191 on Build GRCh38
          UniGene ID:
          Hs.513871
          Hs.369998
          Hs.46835
          Hs.97858
          Hs.201805
          -
          Hs.135997
          Hs.4997
          Hs.145150
          Hs.211079
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          siRNA ID HSS164081
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price Contact Us
          Your Price
          Online offer:
          3062.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CYB5D2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_144611.3 NP_653212.1 1 168
          GenBank mRNA BC051697.1 - 1 80 5190840
          DA346215.1 - 1 145
          Gene Symbol: DBF4B
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_017025152.1 XP_016880641.1 5 2208
          GenBank mRNA AK023149.1 - 5 2291
          Gene Symbol: IAPP
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA J04422.1 - 3 863
          Gene Symbol: KIF1B
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_183416.3 NP_904325.2 21 6582
          Gene Symbol: LAMC3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA AL133595.1 - 6 3007
          Gene Symbol: LINC00269
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_103715.1 5 685
          GenBank mRNA AK090929.1 - 5 685
          Gene Symbol: PCYT1A
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA AK001567.1 - 1 477
          Gene Symbol: RBM23
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA AK125593.1 - 11 2291
          AL834198.1 - 11 2290
          Gene Symbol: SP110
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_005246525.3 XP_005246582.1 20 5439
          XM_006712489.3 XP_006712552.1 18 5226
          XM_011511088.2 XP_011509390.1 20 5517
          XM_011511089.2 XP_011509391.1 19 5372
          XM_011511090.2 XP_011509392.1 19 5367
          XM_011511091.2 XP_011509393.1 19 5298
          XM_011511092.2 XP_011509394.1 18 5210
          XM_017003968.1 XP_016859457.1 21 5592
          Gene Symbol: ZNHIT1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA AK092478.1 - 1 246
          AK125123.1 - 2 2672

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          transmembrane signal receptor kinase activator peptide hormone microtubule binding motor protein RNA splicing factor

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of neuron differentiation
          cell cycle
          positive regulation of cell proliferation
          positive regulation of nuclear cell cycle DNA replication
          positive regulation of G2/M transition of mitotic cell cycle
          activation of protein kinase activity
          apoptotic process
          microtubule-based movement
          neuron-neuron synaptic transmission
          neuromuscular synaptic transmission
          anterograde axonal transport
          cytoskeleton-dependent intracellular transport
          mitochondrion transport along microtubule
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          signal transduction
          viral process
          heme binding
          nucleic acid binding
          protein binding
          zinc ion binding
          protein kinase binding
          protein kinase activator activity
          microtubule motor activity
          ATP binding
          microtubule binding
          ATPase activity
          kinesin binding
          DNA binding
          signal transducer activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-2rf75:80/100.66.79.83:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0