Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPATA31C1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145124.1 | NP_001138596.1 | 4 | 2815 | ||
XM_011518701.2 | XP_011517003.1 | 5 | 3453 | |||
XM_011518702.1 | XP_011517004.1 | 4 | 3372 | |||
XM_011518703.1 | XP_011517005.1 | 4 | 3252 |
Gene Symbol: SPATA31C2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166137.1 | NP_001159609.1 | 4 | 2797 | ||
GenBank mRNA | AK302448.1 | - | 4 | 2797 | ||
AL080179.1 | - | 1 | 299 |