This product has been added to your favorites list. Go to
My Favorites
LOADING 
- Gene Symbol:
-
RCC1L
- Gene Name:
-
RCC1 like
- Gene Aliases:
-
WBSCR16
- Chromosome Location:
-
Chr.7: 75027119 - 75073881 on Build GRCh38
- UniGene ID:
-
Hs.529623
- Species:
- Human
- Species Specific ID (Flybase ID):
-
-
siRNA Details
|
Gene Symbol:
RCC1L |
| Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
| RefSeq |
NM_001281441.1 |
NP_001268370.1 |
3, 4
|
738
|
|
| NM_030798.4 |
NP_110425.2 |
3, 4
|
738
|
|
| NM_148842.2 |
NP_683682.1 |
3, 4
|
738
|
|
| GenBank mRNA |
AF410455.1 |
- |
3, 4
|
594
|
|
| AK057870.1 |
- |
|
977
|
|
| AK309117.1 |
- |
3, 4
|
647
|
|
| AL136804.1 |
- |
3, 4
|
645
|
|
| AL536085.3 |
- |
3, 4
|
736
|
|
| BC007823.2 |
- |
3, 4
|
650
|
4304040 |
| BC019008.2 |
- |
3, 4
|
650
|
4330507 |
| BC032712.1 |
- |
3, 4
|
669
|
5495794 |
| BC040695.1 |
- |
3, 4
|
720
|
5785681 |
| BC157844.1 |
- |
1
|
91
|
9057063 |
Back To Top
Related Products
-
Anti-GAPDH, Mouse Monoclonal 6C5
-
Anti-β-Actin, Mouse Monoclonal AC-15
-
TaqMan® Gene Expression Cells-to-CT™
-
Neon™ Transfection System 100 µL Kit
-
Lipofectamine™ RNAiMAX Transfection Reagent
-
Invivofectamine 2.0
-
Lipofectamine™ 2000 Transfection Reagent
-
Neon™ Transfection System