Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBMXL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162536.2 | NP_001156008.1 | 3 | 984 | ||
NM_019610.5 | NP_062556.2 | 2 | 884 | |||
GenBank mRNA | AL832554.1 | - | 2 | 737 | ||
BC012942.1 | - | 2 | 543 | 3875565 |