Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASPDH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024656.2 | NP_001019827.2 | 2 | 236 | ||
NM_001114598.1 | NP_001108070.1 | 3 | 282 | |||
GenBank mRNA | BC066359.1 | - | 2 | 236 | 5302622 | |
BM695973.1 | - | 2 | 194 | |||
BM924033.1 | - | 3 | 282 |