Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other WHSC1 SIRNA Assays ›
          Gene Symbol:
          WHSC1
          Gene Name:
          Wolf-Hirschhorn syndrome candidate 1
          Gene Aliases:
          MMSET, NSD2, REIIBP, TRX5, WHS
          Chromosome Location:
          Chr.4: 1871357 - 1982207 on Build GRCh38
          UniGene ID:
          Hs.113876
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS167044
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: WHSC1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001042424.2 NP_001035889.1 2 726
          NM_007331.1 NP_015627.1 4 1044
          NM_133330.2 NP_579877.1 4 1044
          NM_133331.2 NP_579878.1 3 864
          NM_133334.2 NP_579889.1 1 579
          NM_133335.3 NP_579890.1 2 689
          XM_005248001.3 XP_005248058.1 2 669
          XM_005248002.2 XP_005248059.1 2 765
          XM_005248005.2 XP_005248062.1 2 765
          XM_006713914.3 XP_006713977.1 3 934
          XM_011513557.2 XP_011511859.1 2 605
          GenBank mRNA AF071593.1 - 1 579
          AF071594.1 - 1 579
          AF083386.1 - 4 1044
          AF083387.1 - 3 864
          AF083388.1 - 2 688
          AF083389.1 - 4 1044
          AF083390.1 - 4 1044
          AF083391.1 - 4 1044
          AJ007042.1 - 2 1312
          AK289697.1 - 3 711
          AY694128.1 - 1 561
          BC020545.1 - 2 661
          BC052254.1 - 2 668
          BC141815.1 - 2 759

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          membranous septum morphogenesis
          atrial septum primum morphogenesis
          atrial septum secundum morphogenesis
          double-strand break repair via nonhomologous end joining
          transcription, DNA-templated
          anatomical structure morphogenesis
          histone H3-K36 methylation
          chromatin modification
          histone H4-K20 methylation
          positive regulation of isotype switching to IgA isotypes
          bone development
          regulation of establishment of protein localization
          regulation of double-strand break repair via nonhomologous end joining
          chromatin binding
          protein binding
          zinc ion binding
          histone-lysine N-methyltransferase activity
          histone methyltransferase activity (H4-K20 specific)
          sequence-specific DNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0