Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101929322 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011516748.2 | XP_011515050.1 | 2, 3 | 544 |
Gene Symbol: INTS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033547.3 | NP_291025.3 | 6, 7 | 747 | ||
XM_011545352.2 | XP_011543654.1 | 7, 8 | 916 | |||
XM_011545353.2 | XP_011543655.1 | 6, 7 | 925 | |||
XM_017018560.1 | XP_016874049.1 | 6, 7 | 780 | |||
GenBank mRNA | AK290866.1 | - | 6, 7 | 743 | ||
AL834171.1 | - | 6, 7 | 690 | |||
BC008013.1 | - | 2, 3 | 200 | |||
BC009995.2 | - | 6, 7 | 742 | 4129693 | ||
BC015664.2 | - | 5, 6 | 640 | 4554197 |