Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other STAG3L2 SIRNA Assays ›
          Gene Symbol:
          STAG3L2 STAG3L1 STAG3L3 STAG3L5P STAG3L5P-PVRIG2P-PILRB TRIM74
          Gene Name:
          stromal antigen 3-like 2 (pseudogene)
          stromal antigen 3-like 1 (pseudogene)
          stromal antigen 3-like 3 (pseudogene)
          stromal antigen 3-like 5 pseudogene
          STAG3L5P-PVRIG2P-PILRB readthrough
          tripartite motif containing 74
          Gene Aliases:
          STAG3L1, STAG3L2P, STAG3L3
          STAG3L1P, STAG3L2, STAG3L3
          STAG3L1, STAG3L2, STAG3L3P
          -
          LOC101735302-LOC101752334-PILRB
          TRIM50C
          Chromosome Location:
          Chr.7: 74881990 - 74890612 on Build GRCh38
          Chromosome Location:
          Chr.7: 75359200 - 75367809 on Build GRCh38
          Chromosome Location:
          Chr.7: 72997295 - 73005943 on Build GRCh38
          Chromosome Location:
          Chr.7: 100336079 - 100341328 on Build GRCh38
          Chromosome Location:
          Chr.7: 100336065 - 100367831 on Build GRCh38
          Chromosome Location:
          Chr.7: 72954845 - 73005932 on Build GRCh38
          UniGene ID:
          Hs.632310
          Hs.632310
          Hs.632310
          Hs.521075
          Hs.521075
          Hs.645328
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          -
          -
          siRNA ID HSS167709
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: STAG3L2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_040584.2 2 257
          GenBank mRNA AK303473.1 - 3 211
          AK307228.1 - 3 328
          AK309007.1 - 3 212
          BC002581.2 - 3 199
          BG537494.1 - 2 256
          BI753182.1 - 3 202
          CB956482.1 - 2 236
          Gene Symbol: STAG3L1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_040583.2 3 381
          GenBank mRNA AK289466.1 - 4 350
          AK291820.1 - 1 21
          BG829469.1 - 2 265
          DC419304.1 - 2 359
          Gene Symbol: STAG3L3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_040582.2 2 400
          GenBank mRNA AK303592.1 - 2 61
          AK316130.1 - 2 263
          AY313779.1 - 2 255
          BC066937.1 - 2 133 4826886
          BC108680.1 - 2 267
          BI829811.1 - 2 135
          BM007884.1 - 2 194
          DA505391.1 - 2 272
          DA555040.1 - 2 282
          DC395490.1 - 2 183
          DC405605.1 - 2 382
          DC410426.1 - 2 61
          Gene Symbol: STAG3L5P
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_103720.1 2 174
          GenBank mRNA BM826568.1 - 2 128
          DA306290.1 - 2 174
          Gene Symbol: STAG3L5P-PVRIG2P-PILRB
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_036569.1 2 305
          NR_036570.1 2 305
          GenBank mRNA AK056412.1 - 2 256
          AK310352.1 - 2 109
          AL834317.1 - 2 127
          DB275512.1 - 2 305
          Gene Symbol: TRIM74
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_017012197.1 XP_016867686.1 2 272

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          chromatin/chromatin-binding, or -regulatory protein

          Gene Ontology Categories:

          Process(es)

          zinc ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-gclgf:80/100.66.78.196:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0