Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PMS2P5 SIRNA Assays ›
          Gene Symbol:
          PMS2P5 DTX2P1-UPK3BP1-PMS2P11 LOC441259 PMS2P1 PMS2P2 PMS2P3 PMS2P4 PMS2P6 PMS2P7 PMS2P9 POLR2J2 POLR2J3 POLR2J4 UPK3B
          Gene Name:
          PMS1 homolog 2, mismatch repair system component pseudogene 5
          DTX2P1-UPK3BP1-PMS2P11 readthrough, transcribed pseudogene
          PMS1 homolog 2, mismatch repair system component pseudogene
          PMS1 homolog 2, mismatch repair system component pseudogene 1
          PMS1 homolog 2, mismatch repair system component pseudogene 2
          PMS1 homolog 2, mismatch repair system component pseudogene 3
          PMS1 homolog 2, mismatch repair system component pseudogene 4
          PMS1 homolog 2, mismatch repair system component pseudogene 6
          PMS1 homolog 2, mismatch repair system component pseudogene 7
          PMS1 homolog 2, mismatch repair system component pseudogene 9
          RNA polymerase II subunit J2
          RNA polymerase II subunit J3
          RNA polymerase II subunit J4, pseudogene
          uroplakin 3B
          Gene Aliases:
          PMS2L5, PMS7
          PMS2L11
          -
          PMS2L1, PMS2L13, PMS2L6, PMS2L7, PMS2L8, PMS3, PMS8, PMSR1, PMSR2
          PMS2L2, PMS4
          PMS2L3, PMS2L9, PMS5, PMSR3
          PMS2L4, PMS6
          PMS2L14
          PMS2L15, PMS2LP2, PMSR7
          PMS2L17, PMS2LP1, PMSR5
          HRPB11B, RPB11b1
          POLR2J2, RPB11b1, RPB11b2
          RPB11-phi
          P35, UP3B, UPIIIB
          Chromosome Location:
          Chr.7: 74890768 - 74921138 on Build GRCh38
          Chromosome Location:
          Chr.7: 76980822 - 77023761 on Build GRCh38
          Chromosome Location:
          Chr.7: 75296553 - 75299806 on Build GRCh38
          Chromosome Location:
          Chr.7: 100320640 - 100336307 on Build GRCh38
          Chromosome Location:
          Chr.7: 75343937 - 75358997 on Build GRCh38
          Chromosome Location:
          Chr.7: 75507747 - 75528123 on Build GRCh38
          Chromosome Location:
          Chr.7: 67276131 - 67302442 on Build GRCh38
          Chromosome Location:
          Chr.7: 73093644 - 73096950 on Build GRCh38
          Chromosome Location:
          Chr.7: 73006080 - 73016375 on Build GRCh38
          Chromosome Location:
          Chr.7: 77039480 - 77053038 on Build GRCh38
          Chromosome Location:
          Chr.7: 102636748 - 102671729 on Build GRCh38
          Chromosome Location:
          Chr.7: 102537919 - 102572660 on Build GRCh38
          Chromosome Location:
          Chr.7: 43940895 - 44019194 on Build GRCh38
          Chromosome Location:
          -
          UniGene ID:
          Hs.292996
          Hs.675888
          Hs.292996
          Hs.634244
          Hs.292996
          Hs.659871
          Hs.712714
          Hs.661055
          Hs.292996
          Hs.292996
          Hs.610255
          Hs.610255
          Hs.657028
          Hs.488861
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          siRNA ID HSS167794
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price Contact Us
          Your Price
          Online offer:
          3062.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PMS2P5
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_027775.2 6 620
          NR_027776.2 5 517
          GenBank mRNA AA621085.1 - 2 242
          BF984173.1 - 5 516
          D38436.1 - 6 601
          D38502.1 - 6 601
          Gene Symbol: DTX2P1-UPK3BP1-PMS2P11
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_023383.1 10 1533
          GenBank mRNA U38980.1 - 10 1418
          Gene Symbol: LOC441259
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA BC075816.1 - 6 575
          Gene Symbol: PMS2P1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_003613.1 6 1011
          GenBank mRNA AB017004.1 - 6 1011
          D38435.1 - 6 614
          D38499.1 - 6 614
          Gene Symbol: PMS2P2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA AA282042.1 - 2 233
          Gene Symbol: PMS2P3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_028059.1 9 1405
          GenBank mRNA D38437.1 - 5 553
          D38498.1 - 5 553
          Gene Symbol: PMS2P4
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_022007.2 5 503
          NR_046297.1 5 503
          NR_046299.1 5 503
          GenBank mRNA BC029368.1 - 5 480
          CB141754.1 - 5 468
          D38438.1 - 5 472
          D38500.1 - 5 472
          DA885243.1 - 5 504
          Gene Symbol: PMS2P6
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA AB017005.1 - 2 170
          Gene Symbol: PMS2P7
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_130940.1 6 648
          GenBank mRNA AB017006.1 - 6 741
          Gene Symbol: PMS2P9
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_028058.1 4 908
          GenBank mRNA BM845848.1 - 5 449
          Gene Symbol: POLR2J2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_032959.5 NP_116581.3 4 468
          GenBank mRNA AB209398.1 - 3 1333
          BC056864.1 - 4 358
          BC086857.1 - 3 293 6007730
          Gene Symbol: POLR2J3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001097615.2 NP_001091084.2 4 440
          XM_017012372.1 XP_016867861.1 4 479
          XM_017012373.1 XP_016867862.1 4 479
          GenBank mRNA AJ277740.1 - 3 165
          AK309487.1 - 3 227
          DA574442.1 - 3 228
          DA580738.1 - 4 402
          Gene Symbol: POLR2J4
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_003655.3 4 427
          GenBank mRNA AK296066.1 - 3 230
          BC017250.2 - 3 1329
          BC062722.1 - 4 386
          BC105584.1 - 4 409
          BX537878.1 - 4 382
          BX538054.1 - 3 206
          CD674494.1 - 2 124
          DA147922.1 - 4 427
          Gene Symbol: UPK3B
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_030570.3 NP_085047.1 5 1525
          NM_182683.2 NP_872624.1 6 1360
          NM_182684.2 NP_872625.1 7 1440
          GenBank mRNA AK056760.1 - 6 1168
          AK290096.1 - 5 1428
          AY233462.1 - 7 1273
          BC004304.1 - 5 1349 3625247

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          DNA metabolism protein DNA-directed RNA polymerase RNA metabolism protein nucleic acid metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          mismatch repair
          somatic hypermutation of immunoglobulin genes
          phosphorylation
          regulation of transcription, DNA-templated
          biological_process
          transcription from RNA polymerase II promoter
          transcription, DNA-templated
          negative regulation of gene expression
          single-stranded DNA binding
          ATPase activity
          kinase activity
          molecular_function
          RNA polymerase II activity
          DNA binding
          protein dimerization activity
          DNA-directed RNA polymerase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0