Search
Search

DUSP22| Gene Symbol: DUSP22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286555.1 | NP_001273484.1 | 7 | 993 | ||
| NR_104473.1 | 6 | 943 | ||||
| NR_104474.1 | 6 | 910 | ||||
| NR_104475.1 | 4 | 826 | ||||
| XM_011514757.2 | XP_011513059.1 | 4 | 455 | |||
| GenBank mRNA | AB208997.1 | - | 4 | 346 | ||
| AK000383.1 | - | 3 | 319 | |||
| AK074410.1 | - | 3 | 319 | |||
| AK128760.1 | - | 4 | 1314 | |||
| AK296402.1 | - | 7 | 563 | |||
| AK310653.1 | - | 5 | 1016 | |||
| BX362191.2 | - | 4 | 607 | |||
| BX648953.1 | - | 4 | 4992 | |||
| DA950312.1 | - | 6 | 526 | |||
| DB177174.1 | - | 6 | 447 | |||